Learn More
Eppendorf™ S-4-104 Rotor Adapters
Available in a wide range of configurations to suit a variety of applications.
Brand:  Eppendorf™ 5825738004
Code : A0
Additional Details : Weight : 0.97200kg
 View more versions of this product
Product Code. 13286619
  • £286.00 / Pack of 2
In Stock
to see stock.
Includes: 2 piece set
Description
Description
  • For use with Centrifuge 5810/5810 R
Specifications
Specifications
Rotor Adapter
23 x 2.6/8 mL Tubes
S-4-104 rotor for 5810/5810R centrifuge
2 Pack