Eppendorf™ Research plus™ Fixed-Volume Pipetters

Resistant to chemical, thermal, and mechanical stress

Overview
Brand: Eppendorf™

Manufacturer Part Number: 3121000031

UNSPSC: 41121504

Code: 97

Additional Details:
Additional Details: Weight: 4.98950kg



Product Code. 10652795

Quantity Price
1 £ 125.0 / Each
Estimated Shipment
11-05-2017
Add to basket


Description and Specification

Specification

Volume (Metric) 20μL
Autoclavable Yes
For Use With (Equipment) 20μL Pipette tip
Color Gray
Accuracy ±0.8%
Includes Spring loaded tip cone, ejector, light grey operating button

  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume

Documentation