Learn More
Eppendorf™ Research plus™ Fixed-Volume Pipetters
Resistant to chemical, thermal, and mechanical stress
Brand:  Eppendorf™ 3121000090
Code : 97
Additional Details : Weight : 0.28000kg
 View more versions of this product
Product Code. 10674335
  • £142.80 / Each
Estimated Shipment
16-05-2018
Add to basket
Description
Description
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
Specifications
Specifications
±0.6%
1000μL Pipette tip
Blue
Yes
Spring loaded tip cone, ejector, blue operating button
200μL
Videos
Documentation