Learn More
Eppendorf™ Reference™ 2 Pipet Packs - Pick-a-Pack Offer
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants. Available in packs of three, four and six.
Brand:  Eppendorf™ 4920000903
Code : NEW
Additional Details : Weight : 1.71000kg
 View more versions of this product
Product Code. 15250506
  • £737.10 / 1 set
Estimated Shipment
13-06-2018
Add to basket
Description
Description
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
Specifications
Specifications
Three adjustable-vol. pipettes: 0.5-10μL 10-100μL, 100-1000μL
Reference 2 pipet pack
Videos