Learn More
Eppendorf™ Rotor Adapters for Centrifuge 5427 R
For 1.5/2mL tubes and cryotubes
Brand:  Eppendorf™ 5820769009
Code : 99
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15138364
  • £25.80 / Pack of 4
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Specifications
Specifications
1 Cryotube
Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Rotor Adapter
Set of 4
Cryotubes
Videos