Learn More
Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Brand:  Eppendorf™ 0030072286
Code : 97
Additional Details : Weight : 0.76500kg
 View more versions of this product
Product Code. 11873462
  • £57.64 / Pack of 480
Estimated Shipment
23-05-2018
Add to basket
Description
Description
  • Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
  • Tips are uncoated and free from additives
  • Ultra-homogenous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five-year shelf life
Specifications
Specifications
LoRetention Tip
Blue
7.1cm
50 to 1000μL
Yes
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Videos
Documentation