Learn More
Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Brand:  Eppendorf™ 0030072278
Code : 97
Additional Details : Weight : 0.54332kg
 View more versions of this product
Product Code. 11863462
  • £55.11 / Pack of 480
Estimated Shipment
23-05-2018
Add to basket
Description
Description
  • Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
  • Tips are uncoated and free from additives
  • Ultra-homogenous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five-year shelf life
Specifications
Specifications
LoRetention Tip
Yellow
5.3cm
2 to 100μL
Yes
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Videos
Documentation