Learn More
Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Brand:  Eppendorf™ 0030072260
Code : 97
Additional Details : Weight : 0.57600kg
 View more versions of this product
Product Code. 11853462
  • £56.66 / Pack of 480
Estimated Shipment
14-02-2018
Add to basket
Description
Description
  • Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
  • Tips are uncoated and free from additives
  • Ultra-homogenous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five-year shelf life
Specifications
Specifications
LoRetention Tip
Light Gray
4.6cm
0.5 to 20μL
Yes
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Videos
Documentation