Learn More
Eppendorf™ 96-Well twin.tec™ PCR Plates
Ideal for extreme applications such as multiple heat sealing
Brand:  Eppendorf™ 0030133340
Code : 96
Additional Details : Weight : 0.58700kg
 View more versions of this product
Product Code. 10399727
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Manufactured using in.twin-shot in. technology, combining the surface of virgin polypropylene wells with the rigidity of poloycarbonate plate decks and skirts.
  • Wells are 20% thinner than standard thin-wall tubes for optimum heat transfer to sample
  • Polycarbonate plate decks and skirts provide mechanical stability, simplifying plate handling and ensuring a more secure seal
  • Autoclavable
  • Meet SBS (Society for Biomolecular Screening) footprint recommendations
  • Automation friendly
  • Available in skirted or semi-skirted styles in five colors with clear wells
Specifications
Specifications
Yes
RNase, DNase-free
Ideal for extreme applications such as multiple heat sealing
Non-skirt
Colorless
No
Red
Polypropylene
150μL
Videos