missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelial Membrane Antigen/EMA Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-3, Mouse Monoclonal Antibody.
£99.10 - £276.42
Specifications
Antigen | Epithelial Membrane Antigen/EMA Ab-3 |
---|---|
Clone | E29 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12662207
|
Epredia
MS-741-P0 |
Concentrated | 200μg/mL | 100 μL |
£99.10
0.10mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12602217
|
Epredia
MS-741-P1 |
Concentrated | 200μg/mL | 500 μL |
£180.00
0.50mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12652207
|
Epredia
MS-741-P |
Concentrated | 200μg/mL | 1 mL |
£252.00
1mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12622217
|
Epredia
MS-741-R7 |
Prediluted | — | 7 mL |
£156.00
7mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12682207
|
Epredia
MS-741-PABX |
Concentrated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12672207
|
Epredia
MS-741-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.Host Species: Mouse
Clone: E29
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: RPAP
Immunogen: Delipidated extract of human milk fat globule membranes
Molecular Weight: 265-400 kDa
Positive Control: Breast or Colon CA
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Epithelial Membrane Antigen/EMA Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
E29 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Delipidated extract of human milk fat globule membranes | |
Primary |