missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549814
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total Ephrin A3 / EFNA3 protein.
Ephrin A3 Polyclonal antibody specifically detects Ephrin A3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Ephrin A3 | |
Polyclonal | |
Unconjugated | |
EFNA3 | |
AW494418; EFL2; EFL-2; EFNA3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; ephrin A3; ephrin-A3; Epl3; EPLG3; Lerk3; LERK-3; ligand of eph-related kinase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13638, 170901, 1944 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08545, P52797 | |
EFNA3 | |
Synthesized peptide derived from C-terminal of human EFNA3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |