missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ephrin-A1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | Ephrin-A1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Ephrin-A1 Polyclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.Specifications
Ephrin-A1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
B61, ECKLG, EFL1, EFL-1, EFNA1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EphrinA1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
EFNA1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P52793 | |
1942 | |
Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A1 Asp19-Ser182 Accession # P52793 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Ephrin-A1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Ephrin-A1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-B1, rmEphrin-A5, rmEphrin-A4, rmEphrin-A2, and rmEphrin-B1 is observed. |
For Research Use Only