missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB6 Mouse anti-Human, Mouse, Clone: 465323, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | EphB6 |
---|---|
Clone | 465323 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197 |
Gene ID (Entrez) | 2051 |
Gene Accession No. | O15197 |
Description
EphB6 Monoclonal specifically detects EphB6 in Human, Mouse samples. It is validated for Western Blot.Specifications
EphB6 | |
Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197 | |
O15197 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
465323 | |
2051 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human and mouse EphB6 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) EphB6 is observed, 25% cross-reactivity with recombinant human (rh) EphA10 is observed, and no cross-reactivity with rhEphA1, 2, 5, rhEphB3, 4, rmEphA1, 3, 4, 6, 7, 8, rmEphB2, 3, recombinant rat (rr) EphA5, or rrEphB1 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only