missing translation for 'onlineSavingsMsg'
Learn More

Ephrin B1, clone: 25H11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5300-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15277407

  • £138.68 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The monoclonal antibody 25H11 recognizes mouse Ephrin B1, a 47kDa membrane protein. Ephrin B1 belongs to a family of ephrins composed of type A (linked to the cell membrane through a glycosylphosphatidyl inositol group) or type B (transmembrane) proteins. The type B ephrins consist of 3 members (B1, B2, and B3) each of which contains a cytoplasmic domain containing tyrosine residues that can be phosphorylated. Ephrin B and ephrin B receptors (Eph B) can interact to potentiate bidirectional downstream signaling between cells expressing either or both the ligand and receptor. Ephrins have been shown to play a role in neural crest cell development through both neurogenesis and neural migration, as well as development of kidney and heart cyto-architecture. Overexpression of type B ephrins in cancer cells is associated with increased vascularity and highly invasive tumors.Applications Reported: This 25H11 antibody has been reported for use in immunohistochemical staining of frozen tissue sections and immunoprecipitation.Applications Tested: This 25H11 antibody has been tested immunohistochemistry on frozen mouse kidney tissue sections at less than or equal to 10 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

EphB1 (ephrin B1)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q8CBF3
ELK, EPHT2, FLJ37986, Hek6, NET
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunoprecipitation
25H11
Unconjugated
Ephb1
Purified
Ephb1
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
270190
Videos

For Research Use Only.