missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA8 Recombinant Human Protein
EPHA8 Recombinant Human Protein
Brand: Invitrogen PV3844
Code : 50
Additional Details : Weight : 0.05250kg
Description
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.Specifications
NP_065387 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
76.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
EPHA8, HEK6 | |
EPHA8 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 565-1005 | |
58nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
0.38mg/mL | |
2046 | |
EPHA8 Recombinant Human Protein | |
–80°C | |
1 Each | |
EPHA8 Recombinant Human Protein | |
Ephrin and EPH receptor | |
EPHA8 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.