missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA7 Recombinant Human Protein
EPHA7 Recombinant Human Protein
Brand: Invitrogen PV3689
Code : 50
Additional Details : Weight : 0.05250kg
Description
EPHA7 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA7 is involved in embryonic development.Specifications
NP_004431.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
76.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
EPHA7, HEK11, EHK3 | |
EPHA7 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 0.25mM EGTA, 0.1mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 579-998 | |
110nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
GST Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
0.33mg/mL | |
2045 | |
EPHA7 Recombinant Human Protein | |
–80°C | |
1 Each | |
EPHA7 Recombinant Human Protein | |
Ephrin and EPH receptor | |
EPHA7 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.