missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA7 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | EphA7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
EphA7 Polyclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot.Specifications
EphA7 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Cek11, Ebk, EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, Mdk1, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
EPHA7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q61772 | |
2045 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA7 isoform 1 Ala30-Pro554 (Gly540-Glu544 del) Accession # Q61772 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse EphA7 in direct ELISAs and Western blots. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) EphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, and rmEphA8 is observed. |
For Research Use Only