missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA4 Recombinant Human Protein
EPHA4 Recombinant Human Protein
£434.00 - £1602.00
Specifications
Accession Number | NP_004429.1 |
---|---|
Concentration | 0.46mg/mL |
Molecular Weight (g/mol) | 59.7kDa |
Name | EPHA4 Recombinant Human Protein |
Storage Requirements | –80°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11543487
|
Invitrogen
PV3651 |
10μg |
£434.00
10µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13437397
|
Invitrogen
PR6419A |
100μg |
£1602.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system.Specifications
NP_004429.1 | |
59.7kDa | |
–80°C | |
1 Each | |
EPHA4 Recombinant Human Protein | |
Amino acids 616-887 | |
230nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
GST Tag | |
85% by SDS-PAGE |
0.46mg/mL | |
EPHA4 Recombinant Human Protein | |
For research use only. Not for use in diagnostic procedures | |
EPHA4, HEK8 | |
EPHA4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 12.5mM NaCl, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
EPHA4 Recombinant Human Protein | |
Human | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.