missing translation for 'onlineSavingsMsg'
Learn More

EPHA4 Recombinant Human Protein

EPHA4 Recombinant Human Protein

Brand:  Invitrogen PR6419A

Code : Z1

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 13437397

  • £1602.00 / 100µg
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system.
Specifications

Specifications

NP_004429.1
Protein Assays and Analysis, Kinase Assay
59.7kDa
100μg
For research use only. Not for use in diagnostic procedures
EPHA4, HEK8
EPHA4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 12.5mM NaCl, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Amino acids 616-887
230nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
GST Tag
Frozen
0.46mg/mL
Frozen
EPHA4 Recombinant Human Protein
–80°C
1 Each
EPHA4 Recombinant Human Protein
EPH receptor
EPHA4 Recombinant Human Protein
Human
Baculovirus
85% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.