missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epTIPS™ Standard Pipet Tips

Use these pipette tips for reduced-force tip attachment and ejection with complete sealing. Eppendorf™ epTIPS™ Standard Pipet Tips are ideal for use with Eppendorf pipettes.

Brand:  Eppendorf™ 0030000900

Code : A0

Additional Details : Weight : 0.44800kg

 View more versions of this product

Product Code. 11930537

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
  • epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
  • epT.I.P.S. Standard pipette tips can be autoclaved at 121°C
Specifications

Specifications

Standard Pipette Tips
Videos