Learn More
Eppendorf™ epT.I.P.S.™ PipetteTips Set
Optimally coordinated to Eppendorf pipettes and meet EN ISO 8655 requirements
Brand:  Eppendorf™ 0030073207
Code : 96
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10619331
  • £52.46 / Pack of 480
In Stock 2
Add to basket
Description
Description
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Specifications
Pipettor Tip Set
0.1 to 10μL
Anthracite
Yes
Videos
Documentation