Learn More
Eppendorf™ epT.I.P.S.™ PipetteTips Set
Optimally coordinated to Eppendorf pipettes and meet EN ISO 8655 requirements
Brand:  Eppendorf™ 0030073347
Code : 96
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10197191
  • £45.21 / Pack of 240
In Stock 3
Add to basket
Description
Description
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Specifications
Pipettor Tip Set
Red
5 Trays x 96 Tips
500 to 2500μL
Yes
Videos
Documentation