Learn More
Eppendorf™ epTIPS Box
For storing pipettor tips.
Brand:  Eppendorf™ 0030073045
UNSPSC : 41121607
Code : 96
Additional Details : Weight : 0.25500kg
 View more versions of this product
Product Code. 10621442
  • £30.65 / Pack of 96
Estimated Shipment
23-06-2017
Add to basket
Specifications
Description
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Description & Specifications
Specifications
Pipette Tips
1 Reusable Box x 96 Tips
Documentation Alternative Products