Learn More
Eppendorf™ epTIPS Box
For storing pipettor tips.
Brand:  Eppendorf™ 0030073223
Code : 96
Additional Details : Weight : 0.54000kg
 View more versions of this product
Product Code. 10629331
  • £52.47 / Pack of 480
Estimated Shipment
22-05-2018
Add to basket
Description
Description
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Specifications
Pipette Tips
5 Trays x 96 Tips
Videos
Documentation