missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ EP-tips™ Pipet Reload tips
Components are 100 % recyclable. Eppendorf™ EP-tips™ Pipet Reload tips produce reduced waste as compared with disposable racks.
£88.07 - £88.12
Specifications
Color | Red |
---|---|
Autoclavable | Autoclavable |
Format | Rack |
Product Line | epT.I.P.S. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10117441
|
Eppendorf™
0030073789 |
0.5 to 20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10688045
|
Eppendorf™
0030073614 |
50 to 1250 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
- Racks can be autoclaved
- Also available in PCR clean quality, free of human DNA, DNase, RNase and PCR inhibitors
- 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Red | |
Autoclavable | |
Rack | |
epT.I.P.S. |