Learn More
Eppendorf™ EP-tips™ Pipet Reload tips
Components are 100 % recyclable. Eppendorf™ EP-tips™ Pipet Reload tips produce reduced waste as compared with disposable racks.
Brand:  Eppendorf™ 0030073886
Code : 96
Additional Details : Weight : 1.48500kg
 View more versions of this product
Product Code. 10709932
  • £49.61 / Pack of 480
Estimated Shipment
15-05-2018
Add to basket
Description
Description
  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR clean quality, free of human DNA, DNase, RNase and PCR inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Specifications
Pipette Tips
10 x 48 Tips in Racks
Videos
Documentation