Learn More
Eppendorf™ EP-tips™ Pipet Reload tips
Components are 100 % recyclable. Eppendorf™ EP-tips™ Pipet Reload tips produce reduced waste as compared with disposable racks.
Brand: Eppendorf™ 0030073614
Code : A0
Additional Details : Weight : 1.38600kg
Description
- Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
- Racks can be autoclaved
- Also available in PCR clean quality, free of human DNA, DNase, RNase and PCR inhibitors
- 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
50 to 1250 μL | |
Autoclavable | |
epT.I.P.S. |
Red | |
Rack |