Learn More
Eppendorf™ EP-tips™ LoRetention Reload Pipet tips
Maximum recovery and reproducibility. Eppendorf™ EP-tips™ LoRetention Reload Pipet tips are ideally suited for all applications with samples that contain detergents.
Brand:  Eppendorf™ 0030077601
Code : NEW
Additional Details : Weight : 208.00000kg
 View more versions of this product
Product Code. 15254516
  • £5.66 / 1 set
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Ultrahomogenous surface for maximum reproducibility
  • Ultrahydrophobic surface for minimum sample loss
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five year shelf life
  • Available as ep Dualfilter T.I.P.S.™, as well as PCR* clean and Eppendorf™ Quality
Specifications
Specifications
Refill
Low Retention
Videos