Learn More
Eppendorf™ EP-tips™ LoRetention Reload Pipet tips
Maximum recovery and reproducibility. Eppendorf™ EP-tips™ LoRetention Reload Pipet tips are ideally suited for all applications with samples that contain detergents.
Brand:  Eppendorf™ 0030072014
Code : 96
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 10531664
  • £70.22 / Pack of 960
Estimated Shipment
19-03-2018
Add to basket
Description
Description
  • Ultrahomogenous surface for maximum reproducibility
  • Ultrahydrophobic surface for minimum sample loss
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five year shelf life
  • Available as ep Dualfilter T.I.P.S.™, as well as PCR* clean and Eppendorf™ Quality
Specifications
Specifications
Loretention Reload Pipette Tips
10 Trays x 96 Tips
Videos
Documentation