Learn More
EOMES Monoclonal Antibody (Dan11mag), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 61-4875-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This Dan11mag antibody recognizes Eomesodermin (EOMES), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This Dan11mag antibody has been reported for use in flow cytometric analysis. Applications Tested: This Dan11mag antibody has been tested by intracellular staining and flow cytometric of mouse splenocytes analysis using Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
EOMES Monoclonal antibody specifically detects EOMES in Mouse samples. It is validated for Flow Cytometry.Specifications
EOMES | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O54839 | |
Eomes | |
Affinity chromatography | |
RUO | |
13813 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
Dan11mag | |
PE-eFluor 610 | |
Eomes | |
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.