missing translation for 'onlineSavingsMsg'
Learn More

EOMES Rat anti-Mouse, PE-Cyanine7, Clone: Dan11mag, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-4875-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15598546

  • £92.75 / 25µg

EU Stock 5

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
Specifications

Specifications

EOMES
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54839
Eomesodermin homolog; EOMES; TBR2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
Dan11mag
PE-Cyanine7
Eomes
Liquid
Eomes
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
13813
Videos
Documents

For Research Use Only.