Learn More
EOMES Monoclonal Antibody (21Mags8), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£76.00 - £185.00
Spécification
Antigen | EOMES |
---|---|
Clone | 21Mags8 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
15237397
|
Affymetrix eBioscience
14-4876-82 |
100 μg |
£185.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15227397
|
Affymetrix eBioscience
14-4876-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This 21Mags8 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Note: For detection of mouse Eomes, we recommend 12-4875 and 51-4875 and for detection of human Eomes, we recommend 51-4877 due to the brighter staining intensity of these products compared to clone 21Mags8. Applications Reported: This 21Mags8 antibody has been reported for use in flow cytometric analysis, and immunoblotting (WB). Applications Tested: This 21Mags8 antibody has been tested by western blot analysis of mouse splenocytes. As a starting concentration, this antibody can be used at 2 μg/mL. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.Spécification
EOMES | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
Eomes | |
Primary | |
4° C | |
Eomes |
21Mags8 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
O54839, O95936 | |
13813, 8320 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.