Learn More
Sartorius Entris™ II Advanced Precision Balances
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA2201-1S
1474.41 GBP valid until 2024-12-31
Use promo code "21987" to get your promotional price.
Additional Details : Weight : 5.70000kg
 View more versions of this product
Product Code. 16652454
  • £1620.00 / Each
In Stock
to see stock.
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results by using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
CE Certificate, CSA for Canada
Without Internal Calibration
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤0.9 seconds
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
2200 g
100 mg
Electronic Precision Balance
100/240 V (± 10%)
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products