missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Enterostatin, Pig, rat

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66038.MA

Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15427239

  • £129.60 / 10mg


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Disclaimers:

For Research Use Only


Specifications

Specifications

582.66
Pig, Rat
Solid
RUO
Enterostatin, pig, rat
HPLC
10mg
Val-Pro-Asp-Pro-Arg
-30° to -10°C
Videos
SDS