missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Enteropeptidase/Enterokinase Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Enteropeptidase/Enterokinase |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Enteropeptidase/Enterokinase Polyclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Enteropeptidase/Enterokinase | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTK, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7, PRSS7enteropeptidase, Serine protease 7, TMPRSS15, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
TMPRSS15 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P98073 | |
5651 | |
Mouse myeloma cell line NS0-derived recombinant human Enteropeptidase/Enterokinase Leu41-His1019 Accession # P98073 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Enteropeptidase/Enterokinase in direct ELISAs and Western blots. |
For Research Use Only