missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-7/Alk-SMase Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | ENPP-7/Alk-SMase |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
ENPP-7/Alk-SMase Polyclonal specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ENPP-7/Alk-SMase | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, ENPP7, Intestinal alkaline sphingomyelinase, MGC50179, NPP7, NPP-7 | |
ENPP7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q6UWV6 | |
339221 | |
Mouse myeloma cell line NS0-derived recombinant human ENPP-7/Alk-SMase Ala22-Ser439 Accession # Q6UWV6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ENPP-7/Alk-SMase in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse ENPP-7 is observed, and less than 5% cross-reactivity with recombinant human (rh) ENPP-2 and rhENPP-5 is observed. |
For Research Use Only