missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | ENPP-5 |
---|---|
Clone | 500915 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
ENPP-5 Monoclonal specifically detects ENPP-5 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ENPP-5 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q9UJA9 | |
59084 | |
Mouse myeloma cell line NS0-derived recombinant human ENPP-5 Pro25-Ser430 Accession # Q9UJA9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ENPP-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ENPP-2, rhENPP-7, or recombinant mouse ENPP-7 is observed. |
500915 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, ENPP5, KIAA0879, NPP5, NPP-5 | |
ENPP5 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only