Learn More
Enoximone, Tocris Bioscience™
PDE3 inhibitor
Brand:  Tocris (Bio-Techne) 169350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13429933
  • £502.74 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Inhibitor of type III phosphodiesterase (PDE3). Increases intracellular cyclic AMP (cAMP) concentrations and enhances myocardial contractility. Also induces concentration-dependent vasodilation in vitro.
Specifications
Chemical Identifiers
77671-31-9
248.3
enoximone, perfan, fenoximone, enoximonum latin, enoximona spanish, enoximona, enoximonum, perfane, unii-c7z4iti7l7, enoximone usan:ban:inn
4-methyl-5-(4-methylsulfanylbenzoyl)-1,3-dihydroimidazol-2-one
C12H12N2O2S
ZJKNESGOIKRXQY-UHFFFAOYSA-N
53708
CC1=C(NC(=O)N1)C(=O)C2=CC=C(C=C2)SC
Specifications
50mg
77671-31-9
enoximone, perfan, fenoximone, enoximonum latin, enoximona spanish, enoximona, enoximonum, perfane, unii-c7z4iti7l7, enoximone usan:ban:inn
CC1=C(NC(=O)N1)C(=O)C2=CC=C(C=C2)SC
248.3
248.3
Enoximone
C12H12N2O2S
ZJKNESGOIKRXQY-UHFFFAOYSA-N
4-methyl-5-(4-methylsulfanylbenzoyl)-1,3-dihydroimidazol-2-one
53708
>99%