missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ENO2 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding mature form of human ENO2 (Met 1-Leu 434) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 13219H08E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15256265

  • £317.51 / 50µg


Estimated Shipment
17-05-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
2026
≥90% by SDS-PAGE
Human
<1 EU/μg
ENO2
Other Proteins
ENO2 Recombinant Human Protein
Lyophilized
48.6kd
50μg
Full Length, Recombinant
Enolase 2
Assay Standard
E.coli
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.