missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Engineers workbench, standard steel top, 840mm x 1800mm x 750mm
Brand: Manutan A349767
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Strong versatile workbenches ideal for use in heavy industries, workshops and maintenance. Heavy gauge, all steel, welded 50mm x 50mm x 5mm angle framework construction. Powder coated dove grey finish.- Load capacity: 750kg
- Dimensions (H x W x D): 840mm x 1800mm x 750mm