missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Engineers workbench, standard laminated top, 840mm x 1200mm x 750mm

Brand:  Key Industrial Equipment™ A349781

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12888748

  • £525.02 / Each

Estimated Shipment date
from Supplier 29-08-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Strong versatile workbenches ideal for use in heavy industries, workshops and maintenance. Heavy gauge, all steel, welded 50mm x 50mm x 5mm angle framework construction. Powder coated dove grey finish.
  • Load capacity: 750kg
  • Dimensions (H x W x D): 840mm x 1200mm x 750mm
Videos
Documents