missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Endothall
Click to view available options
Quantity:
20 mg
100 mg
Description
Endothall is used as an inhibitor of PP2A phosphatase activity. It is also used as a defoliant. It is used as a cell adhesion inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 100 mg |
Description | White |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A moderately potent inhibitor of protein phosphatase 2A |
Formula Weight | 186.20g/mol |
Product Type | Signal Transduction Reagent |
Form | Powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'