missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endosialin/CD248/TEM1 Antibody (OTI4B8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72075
This item is not returnable.
View return policy
Description
Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot.
Specifications
| Endosialin/CD248/TEM1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1 | |
| Mouse | |
| 80.7 kDa | |
| 100 μg | |
| 57124 | |
| Human | |
| Lyophilized |
| Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| CD248 | |
| Human recombinant protein fragment corresponding to amino acids 20-300 of human CD248(NP_065137) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction