missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Endokinin C, Human

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66600.MA

Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15428019

  • £296.10 / 10mg


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

1674.94
Human
Solid
RUO
Endokinin C, human
HPLC
10mg
Lys-Lys-Ala-Tyr-Gln-Leu-Glu-His-Thr-Phe-Gln-Gly-Leu-Leu-NH2
-30° to -10°C
Videos
SDS

For Research Use Only