missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ ZHE Hazardous Waste Filtration System Replacement Parts and Accessories: 90mm Diameter

Designated by the US EPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP)

Brand:  EMD Millipore™ XX2702512

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10768051

  • £234.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

2-way Valve, 1/8 in. NPTM
316 Stainless Steel
ZHE Hazardous Waste Filtration System (90 mm)
Videos