Learn More
Merck Millipore Vent Filters for Pure Water Storage Reservoirs
For use with Pure Water Reservoirs
Brand: Merck Millipore TANKMPK02
Additional Details : Weight : 0.10000kg
Description
For use with Pure Water Reservoirs
- Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
Specifications
Polyethylene | |
RiOs™ 5/8/16 system | |
Hydrophobic | |
Vent Filter |