Learn More
EMD Millipore™ UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Brand:  EMD Millipore™ ZLXUVLP01
Code : Z1
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13103260
  • £244.00 / Each
Estimated Shipment
23-07-2018
Add to basket
Description
Description
  • Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
Specifications
Elix™ 3/5/10 water purification system
UV lamp
254nm
Videos