Learn More
EMD Millipore™ UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Brand:  EMD Millipore™ ZFA10UVM1
Code : NEW
Additional Details : Weight : 4.53000kg
 View more versions of this product
Product Code. 12058942
  • £665.00 / Each
Estimated Shipment
15-06-2018
Add to basket
Description
Description
  • Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
Specifications
Milli-Q™ Advantage a10™ TOC monitor
UV lamp
254nm
Videos