Learn More
EMD Millipore™ UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Brand:  EMD Millipore™ ZFRES00UV
Code : X3
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10334862
  • £187.90 / Each
Estimated Shipment
10-07-2018
Add to basket
Description
Description
  • Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
Specifications
ASM Pure water and older SDS reservoir
UV lamp
254nm
Videos