Learn More
EMD Millipore™ UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Brand:  EMD Millipore™ QUVLQSL01
Code : Z1
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10405252
  • £512.00 / Each
Estimated Shipment
02-03-2018
Add to basket
Specifications
Specifications
Milli-Q™ gradient and synthesis model (blue door) water purification system
UV lamp
254/185nm
Videos